Thermo Scientific Chemicals

Lithium dimethylamide, 96%, Thermo Scientific Chemicals

Catalog number: 044145.14
25 g, Each
Thermo Scientific Chemicals

Lithium dimethylamide, 96%, Thermo Scientific Chemicals

Catalog number: 044145.14
25 g, Each
Quantity
Catalog number: 044145.14
also known as 044145-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3585-33-9
IUPAC Name
lithium(1+) dimethylamine hydride
Molecular Formula
C2H8LiN
InChI Key
QHXBFEDDNKOZHB-UHFFFAOYSA-N
SMILES
[H-].[Li+].CNC
Appearance (Color)
White to yellow
Form
Powder and/or chunks
Assay from Supplier's CofA
≥95.0% (titration)

Description

It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text