Thermo Scientific Chemicals

Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 044149.09
10 g, Each
Thermo Scientific Chemicals

Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 044149.09
10 g, Each
Quantity
Catalog number: 044149.09
also known as 044149-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
814-87-9
IUPAC Name
dialuminium(3+) trioxalate
Molecular Formula
C6Al2O12
InChI Key
ZCLVNIZJEKLGFA-UHFFFAOYSA-H
SMILES
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Appearance (Color)
White
Form
Powder or chunks
Assay (unspecified)
≥98.0%
Water Content
Report waters of hydration

Description

Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text