2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%, Thermo Scientific Chemicals
2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number 044150.03
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%, Thermo Scientific Chemicals
Catalog number044150.03
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1898-66-4
IUPAC Name1,1-diphenyl-2-(2,4,6-trinitrophenyl)hydrazine
Molecular FormulaC18H13N5O6
InChI KeyWCBPJVKVIMMEQC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=C(NN(C2=CC=CC=C2)C2=CC=CC=C2)C(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to black
Assay (unspecified)≥90.0% ( Elemental analysis or HPLC)
FormPowder
2,2-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger. It serves as a general antioxidant detector. It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals. Further, it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only