Thermo Scientific Chemicals

Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 044151.30
250 g, Each
Thermo Scientific Chemicals

Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 044151.30
250 g, Each
Quantity
Catalog number: 044151.30
also known as 044151-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-90-5
IUPAC Name
2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
Molecular Formula
C10H14O4
InChI Key
STVZJERGLQHEKB-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCCOC(=O)C(C)=C
Comment
Stabilizer: 90 to 110 ppm MEHQ
Water Content (Karl Fischer Titration)
0.1% Max (at time of analysis)
Assay (GC)
> 97.5%

Description

Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text