Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0), 96%, Thermo Scientific Chemicals

Catalog number: 044157.09
10 g, Each
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0), 96%, Thermo Scientific Chemicals

Catalog number: 044157.09
10 g, Each
Quantity
Catalog number: 044157.09
also known as 044157-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1295-35-8
IUPAC Name
bis((1Z,5Z)-cycloocta-1,5-diene) nickel
Molecular Formula
C16H24Ni
InChI Key
JRTIUDXYIUKIIE-KZUMESAESA-N
SMILES
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Form
crystalline powder
Gravimetric analysis
≥ 95.0%
Appearance (Color)
Yellow to orange

Description

Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. It is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for isomerization and hydrosilyltion of unsaturated compounds
  • Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene
  • Packaged under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text