Tris(dibenzoylmethane)mono(phenanthroline)europium(III), Thermo Scientific Chemicals
Tris(dibenzoylmethane)mono(phenanthroline)europium(III), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(dibenzoylmethane)mono(phenanthroline)europium(III), Thermo Scientific Chemicals

As an organic red light emitter
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number 044175.77
also known as 044175-77
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
0.1 g
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
Tris(dibenzoylmethane)mono(phenanthroline)europium(III), Thermo Scientific Chemicals
Catalog number044175.77
Price (USD)/ Each
103.00
-
Add to cart
Specifications
Chemical Name or MaterialTris (dibenzoylmethane)mono (phenanthroline)europium (III)
CAS17904-83-5
Melting Point172°C to 173°C
Recommended StorageAmbient temperatures
TSCANo
View more
As an organic red light emitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only