Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044181.06
also known as 044181-06
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
Add to cart
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Catalog number044181.06
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Chemical Identifiers
CAS7440-00-8
IUPAC Nameneodymium
Molecular FormulaNd
InChI KeyQEFYFXOXNSNQGX-UHFFFAOYSA-N
SMILES[Nd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only