Thermo Scientific Chemicals

Nickel Chromium wire, Thermo Scientific Chemicals

Catalog number: 044187.H9
500 m, Each
Thermo Scientific Chemicals

Nickel Chromium wire, Thermo Scientific Chemicals

Catalog number: 044187.H9
500 m, Each
Quantity
Catalog number: 044187.H9
also known as 044187-H9
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11106-97-1
IUPAC Name
chromium nickel
Molecular Formula
CrNi
InChI Key
VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES
[Cr].[Ni]
Comment
Ni 80% min
Comment
0.05 mm diameter

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text