Thermo Scientific Chemicals

Borane-d{3}, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 044188.AD
50 mL, Each
Thermo Scientific Chemicals

Borane-d{3}, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 044188.AD
50 mL, Each
Quantity
Catalog number: 044188.AD
also known as 044188-AD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13763-62-7
IUPAC Name
(²H₃)borane
Molecular Formula
BD3
InChI Key
UORVGPXVDQYIDP-FIBGUPNXSA-N
SMILES
[2H]B([2H])[2H]
Form
Liquid
Comment
Stabilizer: NaBD4 (0.005M)
Comment
2H NMR for Concentration: 0.8-1.3M
Comment
GC/MS for Isotopic Enrichment: ≥98.0%
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text