Thermo Scientific Chemicals

Fullerene, nanotube, multi-walled, 20-50 nm OD, 5-20 micron long, Thermo Scientific Chemicals

Catalog number: 044192.03
1 g, Each
Thermo Scientific Chemicals

Fullerene, nanotube, multi-walled, 20-50 nm OD, 5-20 micron long, Thermo Scientific Chemicals

Catalog number: 044192.03
1 g, Each
Quantity
Catalog number: 044192.03
also known as 044192-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
308068-56-6
Form
Powder
Assay from Supplier's CofA
≥94.0%
Diameter
Report (nm)
Length
Report (nm)

Description

Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text