Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 044199.22
also known as 044199-22
Price (USD)/ Each
642.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
642.00
Add to cart
Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number044199.22
Price (USD)/ Each
642.00
-
Add to cart
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Employed as adsorbents, functional fluids (closed systems) and intermediates. Used as an operating medium for air-conditioning and industrial drying system. It is also used in manufacturing pharmaceuticals and alkylation process. It is used as brazing and welding fluxes. Used for reconstitution of brines, as swelling agent for proteins, as electrolyte component in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only