Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals

Wetting and solubilizing agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044203.22
also known as 044203-22
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
Add to cart
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Catalog number044203.22
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
-
Add to cart
Chemical Identifiers
CAS577-11-7
IUPAC Namesodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
Molecular FormulaC20H37NaO7S
InChI KeyAPSBXTVYXVQYAB-UHFFFAOYNA-M
SMILES[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤100 (50% solid in 1:1 alcohol/water)
FormWaxy solid
Acid Value≤2.5 (on solids basis)
Iodine value≤0.25 (on solids basis)
Appearance (Color)White
View more
Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only