Thermo Scientific Chemicals

Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals

Catalog number: 044203.36
500 g, Each
Thermo Scientific Chemicals

Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals

Catalog number: 044203.36
500 g, Each
Quantity
Catalog number: 044203.36
also known as 044203-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
577-11-7
IUPAC Name
sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
Molecular Formula
C20H37NaO7S
InChI Key
APSBXTVYXVQYAB-UHFFFAOYNA-M
SMILES
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
Color (APHA)
≤100 (50% solid in 1:1 alcohol/water)
Form
Waxy solid
Acid Value
≤2.5 (on solids basis)
Iodine value
≤0.25 (on solids basis)
Appearance (Color)
White

Description

Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text