Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals
Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 044222.22
also known as 044222-22
Price (USD)/ Each
1,395.65
Online exclusive
1551.00 
Save 155.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
1,395.65
Online exclusive
1551.00 
Save 155.35 (10%)
Add to cart
Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals
Catalog number044222.22
Price (USD)/ Each
1,395.65
Online exclusive
1551.00 
Save 155.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPlatinum
Name Note5% on alumina powder
CAS7440-6-6
Health Hazard 1H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
AppearancePowder
View more
Platinum on Alumina (aluminum oxide) is catalyst material with applications in synthetic chemistry such as hydrogenation of functional groups. catalyst is recommended for selective hydrogenation reactions. The particle size of the catalyst is ideal for allowing fast separation from the reaction mixture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only