Thermo Scientific Chemicals

Zinc bromide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044223.18
50 g, Each
Thermo Scientific Chemicals

Zinc bromide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044223.18
50 g, Each
Quantity
Catalog number: 044223.18
also known as 044223-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7699-45-8
IUPAC Name
zinc(2+) dibromide
Molecular Formula
Br2Zn
InChI Key
VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES
[Zn++].[Br-].[Br-]
Form
Powder
Total Metal Impurities
0.001% max.

Description

Mainly used in ocean wells as complete liquid, stationary liquid, amending liquid. It is also an important material in medical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text