Thermo Scientific Chemicals

Lithium perchlorate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 044225.09
10 g, Each
Thermo Scientific Chemicals

Lithium perchlorate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 044225.09
10 g, Each
Quantity
Catalog number: 044225.09
also known as 044225-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-03-9
IUPAC Name
lithium(1+) trihydrate perchlorate
Molecular Formula
ClH6LiO7
InChI Key
JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES
[Li+].O.O.O.[O-][Cl](=O)(=O)=O
Appearance (Color)
White
Assay from Supplier's CofA
≥98.5 to ≤101.5% (Titration)
Impurity content
≤200 ppm Sulfate (SO4)
Water Content (Karl Fischer Titration)
≤1.0%
Chloride content
≤50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text