Thermo Scientific Chemicals

Zinc bromide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044230.22
100 g, Each
Thermo Scientific Chemicals

Zinc bromide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 044230.22
100 g, Each
Quantity
Catalog number: 044230.22
also known as 044230-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
299465-28-4
IUPAC Name
zinc(2+) dibromide
Molecular Formula
Br2Zn
InChI Key
VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES
[Zn++].[Br-].[Br-]
Total Metal Impurities
0.1% max.

Description

Zinc bromide is an optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds. Zinc bromide is employed as a lewis acid in organic chemistry. Has been used as a transparent shield against radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text