Thermo Scientific Chemicals

Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals

Catalog number: 044231.06
5 g, Each
Thermo Scientific Chemicals

Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals

Catalog number: 044231.06
5 g, Each
Quantity
Catalog number: 044231.06
also known as 044231-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51777-38-9
IUPAC Name
2,2,6,6-tetramethyl-4,4-bis({2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}oxy)-3,5-dioxa-2,4,6-trisilaheptane
Molecular Formula
C24H72O10Si11
InChI Key
VLEKPQBXDHFSQO-UHFFFAOYSA-N
SMILES
C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Assay (HPLC)
> 97.0%

Description

Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text