Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
Catalog number 044231.22
also known as 044231-22
Price (USD)/ Each
890.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
890.00
Add to cart
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Catalog number044231.22
Price (USD)/ Each
890.00
-
Add to cart
Chemical Identifiers
CAS51777-38-9
IUPAC Name2,2,6,6-tetramethyl-4,4-bis({2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}oxy)-3,5-dioxa-2,4,6-trisilaheptane
Molecular FormulaC24H72O10Si11
InChI KeyVLEKPQBXDHFSQO-UHFFFAOYSA-N
SMILESC[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)> 97.0%
Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only