Chemical Name or MaterialPalladium nitrate
Health Hazard 1H290-H314
Health Hazard 3P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 8; Packaging Group: III
View more