Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%, Thermo Scientific Chemicals
Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr &lt;1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044244.14
also known as 044244-14
Price (USD)/ Each
975.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1148.00 
Save 172.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
975.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1148.00 
Save 172.35 (15%)
Add to cart
Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%, Thermo Scientific Chemicals
Catalog number044244.14
Price (USD)/ Each
975.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1148.00 
Save 172.35 (15%)
-
Add to cart
Chemical Identifiers
CAS15823-43-5
IUPAC Namehafnium sulfate
Molecular FormulaHfO4S
InChI KeyKOELDGKLEFTHGN-UHFFFAOYSA-L
SMILES[Hf].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentZr<1%
Total Metal Impurities excluding Zr0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only