Thermo Scientific Chemicals

Bicinchoninic acid disodium salt, Thermo Scientific Chemicals

Catalog number: 044247.04
2 g, Each
Thermo Scientific Chemicals

Bicinchoninic acid disodium salt, Thermo Scientific Chemicals

Catalog number: 044247.04
2 g, Each
Quantity
Catalog number: 044247.04
also known as 044247-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
979-88-4
IUPAC Name
disodium [2,2'-biquinoline]-4,4'-dicarboxylate
Molecular Formula
C20H10N2Na2O4
InChI Key
AUPXFICLXPLHBB-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
Appearance (Color)
White to pale cream to yellow
Form
Powder and chunks
Elemental Analysis
C > 59% H > 2.46% N> 6.84%

Description

For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text