Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bicinchoninic acid disodium salt, Thermo Scientific Chemicals

For determination of Cu and protein assays
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 044247.09
also known as 044247-09
Price (USD)/ Each
290.65
Online exclusive
323.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
290.65
Online exclusive
323.00 
Save 32.35 (10%)
Add to cart
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Catalog number044247.09
Price (USD)/ Each
290.65
Online exclusive
323.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS979-88-4
IUPAC Namedisodium [2,2'-biquinoline]-4,4'-dicarboxylate
Molecular FormulaC20H10N2Na2O4
InChI KeyAUPXFICLXPLHBB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to yellow
FormPowder and chunks
Elemental AnalysisC > 59% H > 2.46% N> 6.84%
For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only