Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number 044263.A7
also known as 044263-A7
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
Add to cart
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number044263.A7
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. It plays an important role in textiles as a UV-absorber. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only