Chemical Name or MaterialNon Metals plasma standard solution
Name NoteAs, B, P, S, Se, Si, Te at 100μg/mL
Health Hazard 1Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Quantity100 mL
Health Hazard 2May be corrosive to metals, Causes severe skin burns and eye damage
View more