Chemical Name or MaterialSemi Metals, plasma standard solution
Name NoteAs, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl at 100μg/mL
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Quantity100 mL
Health Hazard 2May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
View more