Thermo Scientific Chemicals

Tris(2-phenylpyridine)iridium(III), Thermo Scientific Chemicals

Catalog number: 044282.77
0.1 g, Each
Thermo Scientific Chemicals

Tris(2-phenylpyridine)iridium(III), Thermo Scientific Chemicals

Catalog number: 044282.77
0.1 g, Each
Quantity
Catalog number: 044282.77
also known as 044282-77
Price (USD)
Have Questions?

Chemical Identifiers

CAS
693794-98-8
IUPAC Name
tris(2-(pyridin-2-yl)benzen-1-ide) iridium
Molecular Formula
C33H24IrN3
InChI Key
JAZWLEYGTKXZCJ-UHFFFAOYSA-N
SMILES
[Ir].C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1
Form
powder
lambda-max (UV-Vis)
Absorption max: 350 to 400 nm
lambda-max (UV-Vis)
Emission max: 500 to 520 nm
Appearance (Color)
Yellow to green

Description

As a green light emitting material

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text