Thermo Scientific Chemicals

n-Octadecane, tech., 90%, Thermo Scientific Chemicals

Catalog number: 044301.A7
5 kg, Each
Thermo Scientific Chemicals

n-Octadecane, tech., 90%, Thermo Scientific Chemicals

Catalog number: 044301.A7
5 kg, Each
Quantity
Catalog number: 044301.A7
also known as 044301-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-45-3
IUPAC Name
octadecane
Molecular Formula
C18H38
InChI Key
RZJRJXONCZWCBN-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC
Assay from Supplier's CofA
≥88.0%
Form
Fused solid or melt

Description

n-octadecane is used as a solvent, a lubricant, transformer oil and an anti-corrosion agent. It is also used in paraffin and as a chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text