Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals

Maximum level of impurities: Other halogens (as Cl) 0.02%, Mn 3ppm
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 044309.36
also known as 044309-36
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
Add to cart
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Catalog number044309.36
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7790-28-5
IUPAC Namesodium periodate
Molecular FormulaINaO4
InChI KeyJQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentMn 3ppm
Impurity contentOther halogens (as Cl) 0.02%
Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only