Thermo Scientific Chemicals

Aluminum chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044313.14
25 g, Each
Thermo Scientific Chemicals

Aluminum chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044313.14
25 g, Each
Quantity
Catalog number: 044313.14
also known as 044313-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-70-0
IUPAC Name
aluminium(3+) trichloride
Molecular Formula
AlCl3
InChI Key
VSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES
[Al+3].[Cl-].[Cl-].[Cl-]
Form
Powder
Appearance (Color)
White
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.

Description

Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text