Lead(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Lead(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Lead(II) Iodide, Ultra Dry, I2Pb, CAS Number-10101-63-0, blei ii-iodid, diiodo-, e2-plumbane, hsdb 636, lead diiodide, lead ii iodide, lead iodide pbi2, pbi2, plumbous iodide, plumbum jodatum, 100g, 99.999% (Metals basis), 954 deg.C, Yellow, 6.16, DOT Class: 6.1, Packaging Group: III, 233-256-9, 461
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044314.14
also known as 044314-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10101-63-0
IUPAC Nameλ²-lead(2+) diiodide
Molecular FormulaI2Pb
InChI KeyRQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES[I-].[I-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)yellow, yellow-orange or orange-brown
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only