Thermo Scientific Chemicals

Lead(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044314.22
100 g, Each
Thermo Scientific Chemicals

Lead(II) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044314.22
100 g, Each
Quantity
Catalog number: 044314.22
also known as 044314-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10101-63-0
IUPAC Name
λ²-lead(2+) diiodide
Molecular Formula
I2Pb
InChI Key
RQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES
[I-].[I-].[Pb++]
Appearance (Color)
yellow, yellow-orange or orange-brown
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.001% max.
Particle Size
-10 Mesh

Description

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text