Thermo Scientific Chemicals

Zinc perchlorate hexahydrate, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 044315.18
50 g, Each
Thermo Scientific Chemicals

Zinc perchlorate hexahydrate, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 044315.18
50 g, Each
Quantity
Catalog number: 044315.18
also known as 044315-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-64-6
IUPAC Name
zinc(2+) hexahydrate diperchlorate
Molecular Formula
Cl2H12O14Zn
InChI Key
PADPILQDYPIHQQ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Zn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Total Metal Impurities
0.003% max.

Description

Used as intermediates. A synthetic reagent for proteomics research. Zinc(II) perchlorate hexahydrate is a highly efficient catalyst for the opening of epoxides by amines affording 2-amino alcohols in high yields under solvent-free conditions and with excellent chemo-, regio-, and stereoselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text