Thermo Scientific Chemicals

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044318.18
50 g, Each
Thermo Scientific Chemicals

Manganese(II) perchlorate hexahydrate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044318.18
50 g, Each
Quantity
Catalog number: 044318.18
also known as 044318-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15364-94-0
IUPAC Name
manganese(2+) hexahydrate diperchlorate
Molecular Formula
Cl2H12MnO14
InChI Key
AIFGVOMWPFMOCN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Total Metal Impurities
0.005% max.

Description

Manganese(II) perchlorate hexahydrate, is a good oxidizing agent and also acts as an intermediates in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text