Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number 044335.G5
also known as 044335-G5
Price (USD)/ Each
601.00
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
601.00
Add to cart
Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9998% (metals basis), Thermo Scientific Chemicals
Catalog number044335.G5
Price (USD)/ Each
601.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
Assay from Supplier's CofA≥99.9999% (metals basis)
FormWire, 2.0mm (0.08in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only