Thermo Scientific Chemicals

Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Catalog number: 044338.06
5 g, Each
Thermo Scientific Chemicals

Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Catalog number: 044338.06
5 g, Each
Quantity
Catalog number: 044338.06
also known as 044338-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-18-8
IUPAC Name
ruthenium
Molecular Formula
Ru
InChI Key
KJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES
[Ru]
Assay from Supplier's CofA
Ruthenium, Ru: 4.3-5.5% (dry weight basis)
Form
Powder
Residual water
45-60%
Appearance (Color)
Black

Description

Ruthenium, 5% on activated carbon powde is used in solar cells, which turn light energy into electrical energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text