Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 044338.14
also known as 044338-14
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
Add to cart
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number044338.14
Price (USD)/ Each
148.65
Online exclusive
165.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofARuthenium, Ru: 4.3-5.5% (dry weight basis)
FormPowder
Residual water45-60%
Appearance (Color)Black

Ruthenium, 5% on activated carbon powde is used in solar cells, which turn light energy into electrical energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only