Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044345.04
2 g, Each
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044345.04
2 g, Each
Quantity
Catalog number: 044345.04
also known as 044345-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6147-53-1
IUPAC Name
λ²-cobalt(2+) diacetate tetrahydrate
Molecular Formula
C4H14CoO8
InChI Key
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.001% max.

Description

Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text