Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 044345.18
also known as 044345-18
Price (USD)/ Each
1,186.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
1,186.00
Add to cart
Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number044345.18
Price (USD)/ Each
1,186.00
-
Add to cart
Chemical Identifiers
CAS6147-53-1
IUPAC Nameλ²-cobalt(2+) diacetate tetrahydrate
Molecular FormulaC4H14CoO8
InChI KeyZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILESO.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only