Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals
Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 044347.14
also known as 044347-14
Price (USD)/ Each
623.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
623.00
Add to cart
Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals
Catalog number044347.14
Price (USD)/ Each
623.00
-
Add to cart
Chemical Identifiers
CAS1308-96-9
IUPAC Namedieuropium(3+) trioxidandiide
Molecular FormulaEu2O3
InChI KeyAEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Eu+3].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.004% max.
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes, red or blue phosphor in television sets. It shows an useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only