Thermo Scientific Chemicals

Iodine monochloride, ACS, Thermo Scientific Chemicals

Catalog number: 044348.36
500 g, Each
Thermo Scientific Chemicals

Iodine monochloride, ACS, Thermo Scientific Chemicals

Catalog number: 044348.36
500 g, Each
Quantity
Catalog number: 044348.36
also known as 044348-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-99-0
IUPAC Name
iodochlorane
Molecular Formula
ClI
InChI Key
QZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILES
ClI
Comment
Assay I/Cl ratio of the Wijs solution prepared from the reagent must be 1.10±0.1
Insoluble material
0.005%

Description

Iodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text