Thermo Scientific Chemicals

Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog number: 044349.22
100 g, Each
Thermo Scientific Chemicals

Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog number: 044349.22
100 g, Each
Quantity
Catalog number: 044349.22
also known as 044349-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-82-8
IUPAC Name
indium(3+) trichloride
Molecular Formula
Cl3In
InChI Key
PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[In+3]
Residual water
< 100 ppm
Appearance (Color)
White
Form
Powder
Total Metal Impurities
0.001% max.

Description

Indium(III) chloride is used as catalyst for aqueous organic reactions including C-C bond formation, aldol reactions, and reductions and for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text