Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044352.14
25 g, Each
Thermo Scientific Chemicals

Lithium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 044352.14
25 g, Each
Quantity
Catalog number: 044352.14
also known as 044352-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-48-7
IUPAC Name
dilithium(1+) sulfate
Molecular Formula
Li2O4S
InChI Key
INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]S([O-])(=O)=O
Total Metal Impurities
0.01% max.

Description

Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text