Thermo Scientific Chemicals

Cobalt(II) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044354.06
5 g, Each
Thermo Scientific Chemicals

Cobalt(II) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 044354.06
5 g, Each
Quantity
Catalog number: 044354.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1307-96-6
IUPAC Name
oxocobalt
Molecular Formula
CoO
InChI Key
IVMYJDGYRUAWML-UHFFFAOYSA-N
SMILES
O=[Co]
Form
Powder
Assay (special)
?2.5 % of Theoretical value (Co)
Total Metal Impurities
≤0.005% (50 ppm)
Appearance (Color)
Green-brown

Description

Cobalt(II) oxide is widely used in the ceramic industry. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text