Platinum, 5% on graphite, Type 286, Thermo Scientific Chemicals
Platinum, 5% on graphite, Type 286, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, 5% on graphite, Type 286, Thermo Scientific Chemicals

Used as a material for gas sensors.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044365.06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentPlatinum content: 4.5-5.5% w/w (analysis of dried sample)
Loss on Drying≤2.0%
Appearance (Color)Grey

Platinum, 5% on graphite, Type 286 is used as a material for gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only