Bis(cyclopentadienyl)zirconium dihydride, 98%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium dihydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)zirconium dihydride, 98%, Thermo Scientific Chemicals

Hydrogenation catalyst for alkenes and alkynes
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044377.03
also known as 044377-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBis (cyclopentadienyl)zirconium dihydride
CAS37342-98-6
Health Hazard 1H228-H261-H314-H500
Health Hazard 2GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
View more
Bis(cyclopentadienyl)zirconium dihydride, is used as a pharmaceutical intermediate and in chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only