Silica gel desiccant, indicating, -6+16 mesh granules, Thermo Scientific Chemicals
Silica gel desiccant, indicating, -6+16 mesh granules, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel desiccant, indicating, -6+16 mesh granules, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number 044389.A1
also known as 044389-A1
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
Silica gel desiccant, indicating, -6+16 mesh granules, Thermo Scientific Chemicals
Catalog number044389.A1
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-6+16 Mesh
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only