Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals
Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals

Formation of amide groups using Boc strategy, as a scavenger resin
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 044392.14
also known as 044392-14
Price (USD)/ Each
207.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
207.00
Add to cart
Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals
Catalog number044392.14
Price (USD)/ Each
207.00
-
Add to cart
Specifications
Chemical Name or MaterialPoly (styrene-divinylbenzene)
Name NoteAminomethylated, 1% cross-linked
CAS89551-24-6
Physical FormPowder
Recommended StorageAmbient temperatures
View more

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh is used in the formation of amide groups using Boc strategy. as a scavenger resin. C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only