Thermo Scientific Chemicals

2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g, Thermo Scientific Chemicals

Catalog number: 044407.09
10 g, Each
Thermo Scientific Chemicals

2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g, Thermo Scientific Chemicals

Catalog number: 044407.09
10 g, Each
Quantity
Catalog number: 044407.09
also known as 044407-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42074-68-0
IUPAC Name
1-chloro-2-(chlorodiphenylmethyl)benzene
Molecular Formula
C19H14Cl2
InChI Key
JFLSOKIMYBSASW-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1C(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Off-white to yellow to orange
Form
100 to 200 mesh beads
Assay (unspecified)
1% cross linked
Assay (unspecified)
substitution level 1.0 to 1.4 mmol/g

Description

2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text