Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals
Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 044409.22
also known as 044409-22
Price (USD)/ Each
64.65
Online exclusive
71.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.90 
Save 7.25 (10%)
Add to cart
Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals
Catalog number044409.22
Price (USD)/ Each
64.65
Online exclusive
71.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentNa 0.01%
Impurity contentPb 0.01%
Impurity contentSO₄ P.T.
Solution Testin hydrochloric acid P.T.
Assay (unspecified)98.0-103.0% Cl₂Sn.2H2O
View more
Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only